Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 18 - 19 | 2107 | 147 | |
| GenBank mRNA | AF094519.1 | - | 18 - 19 | 2107 | 147 | |
| AK138694.1 | - | 18 - 19 | 2137 | 147 | ||
| AK161737.1 | - | 17 - 18 | 2125 | 147 | ||
| AK166882.1 | - | 18 - 19 | 2163 | 147 | ||
| AK220493.1 | - | 18 - 19 | 2171 | 147 | ||
| BC085191.1 | - | 18 - 19 | 2217 | 30693873 | 147 | |
| BC086779.1 | - | 18 - 19 | 2182 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map