Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 19 - 20 | 2201 | 123 | |
| GenBank mRNA | AF094519.1 | - | 19 - 20 | 2201 | 123 | |
| AK138694.1 | - | 19 - 20 | 2231 | 123 | ||
| AK161737.1 | - | 18 - 19 | 2219 | 123 | ||
| AK166882.1 | - | 19 - 20 | 2257 | 123 | ||
| AK220493.1 | - | 19 - 20 | 2265 | 123 | ||
| BC028920.1 | - | 12 - 13 | 1502 | 4008236 | 123 | |
| BC085191.1 | - | 19 - 20 | 2311 | 30693873 | 123 | |
| BC086779.1 | - | 19 - 20 | 2276 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map