Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 7 - 8 | 703 | 110 | |
| GenBank mRNA | AF094519.1 | - | 7 - 8 | 703 | 110 | |
| AK137073.1 | - | 6 - 7 | 792 | 110 | ||
| AK138694.1 | - | 7 - 8 | 733 | 110 | ||
| AK161737.1 | - | 6 - 7 | 721 | 110 | ||
| AK166882.1 | - | 7 - 8 | 759 | 110 | ||
| AK220493.1 | - | 7 - 8 | 767 | 110 | ||
| BC003250.1 | - | 6 - 7 | 737 | 110 | ||
| BC085191.1 | - | 7 - 8 | 813 | 30693873 | 110 | |
| BC086779.1 | - | 7 - 8 | 778 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map