Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 8 - 9 | 842 | 128 | |
| GenBank mRNA | AF094519.1 | - | 8 - 9 | 842 | 128 | |
| AK137073.1 | - | 7 - 8 | 931 | 128 | ||
| AK138694.1 | - | 8 - 9 | 872 | 128 | ||
| AK161737.1 | - | 7 - 8 | 860 | 128 | ||
| AK166882.1 | - | 8 - 9 | 898 | 128 | ||
| AK220493.1 | - | 8 - 9 | 906 | 128 | ||
| BC003250.1 | - | 7 - 8 | 876 | 128 | ||
| BC028920.1 | - | 1 - 2 | 143 | 4008236 | 128 | |
| BC085191.1 | - | 8 - 9 | 952 | 30693873 | 128 | |
| BC086779.1 | - | 8 - 9 | 917 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map