Assay Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ251508.1 | - | 106 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ251508.1 transcripts.
Set Membership: |
|

Zfand6
Genomic Map