Assay Details
| Gene Symbol: Cth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145953.2 | NP_666065.1 | 3 - 4 | 381 | 71 | |
| GenBank mRNA | AK002480.1 | - | 3 - 4 | 365 | 71 | |
| AY083352.1 | - | 3 - 4 | 381 | 71 | ||
| BC019483.1 | - | 3 - 4 | 367 | 4235178 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cth
Genomic Map