Assay Details
| Gene Symbol: Cth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145953.2 | NP_666065.1 | 9 - 10 | 1036 | 80 | |
| GenBank mRNA | AY083352.1 | - | 9 - 10 | 1036 | 80 | |
| BC019483.1 | - | 9 - 10 | 1022 | 4235178 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cth
Genomic Map