Assay Details
Gene Symbol: Fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029330.2 | NP_083606.2 | 3 - 4 | 346 | 94 | |
GenBank mRNA | AJ276067.2 | - | 3 - 4 | 333 | 94 | |
AK028640.1 | - | 3 - 4 | 343 | 94 | ||
AK045369.1 | - | 3 - 4 | 347 | 94 | ||
BC110551.1 | - | 3 - 4 | 346 | 40047377 | 94 | |
BC110552.1 | - | 3 - 4 | 343 | 40047378 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22666539 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22666538 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P229886 | AK045369.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P229886 | AK045369.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109541 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102474 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179066 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P229886 | AK045369.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P229886 | AK045369.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P233927 | AK030969.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P229886 | AK045369.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P229886 | AK045369.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P682020 | BC110552.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P682020 | AK045369.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P682020 | BC110551.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P682020 | AJ276067.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P682020 | AK006023.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179066 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05129988 | - | - |
Whole Mouse Genome G4122A | A_52_P233927 | AK030969.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P682020 | BC110552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P682020 | AK045369.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P682020 | BC110551.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P682020 | AJ276067.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P682020 | AK006023.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P229886 | AK045369.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P229886 | AK045369.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P229886 | AJ276067.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P229886 | NM_029330.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179066 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03022495 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02788211 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02788212 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02788214 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22666538 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22666539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22666450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02378106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03006109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02347973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02565847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03023865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03065537 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03065538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03065546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03065530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02391337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22666472 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10503013 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10503015 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10503012 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10503014 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10503016 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429519_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10503010 | - | - |
Mouse Genome 430 2.0 Array | 1429519_at | - | - |
Mouse Genome 430 2.0 Array | 1429519_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75540_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75540_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300024974.mm.1 | NM_029330.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024974.mm.1 | NM_029330.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024974.mm.1 | TC0300003182.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300024976.mm.1 | NM_029330.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024976.mm.1 | NM_029330.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024976.mm.1 | TC0300003182.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300024973.mm.1 | NM_029330.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024973.mm.1 | NM_029330.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024973.mm.1 | TC0300003182.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429519_PM_at | - | - |
Mouse Expression Set 430 | 1429519_at | - | - |
Murine Genome U74v2 Set | 165709_at | - | - |
Murine Genome U74v2 Set | 134137_at | - | - |
Murine Genome U74v2 Set | 165709_PM_at | - | - |
Murine Genome U74v2 Set | 134137_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429519_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429519_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 372512 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5390273 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2637156 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2637156 | - | - |
