Assay Details
| Gene Symbol: Nexn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199465.2 | NP_955759.2 | 5 - 6 | 460 | 98 | |
| GenBank mRNA | AK136097.1 | - | 5 - 6 | 586 | 98 | |
| AK136102.1 | - | 5 - 6 | 388 | 98 | ||
| AK163414.1 | - | 5 - 6 | 460 | 98 | ||
| AK163848.1 | - | 4 - 5 | 374 | 98 | ||
| BC052878.1 | - | 5 - 6 | 407 | 30046316 | 98 | |
| HQ179982.1 | - | 5 - 6 | 494 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nexn
Genomic Map