Assay Details
| Gene Symbol: Zzz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132612.1 | - | 1 - 2 | 134 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 33 | Usp33 |
9830169D19Rik, AA409780, Vdu1 |
-26.4kb | Sense | Chr. 3 - 152346478 - 152393617 on NCBI Build GRCm38 |
| Downstream | adenylate kinase 5 | Ak5 | 15.9kb | Anti-sense | Chr. 3 - 152462815 - 152668376 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Zzz3
Genomic Map