Assay Details
| Gene Symbol: Erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175176.3 | NP_780385.1 | 3 - 4 | 593 | 77 | |
| GenBank mRNA | AK076632.1 | - | 3 - 4 | 594 | 77 | |
| BC138236.1 | - | 3 - 4 | 513 | 8861255 | 77 | |
| BC145840.1 | - | 3 - 4 | 513 | 40131143 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erich3
Genomic Map