Assay Details
| Gene Symbol: Slc44a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081263.1 | NP_001074732.1 | 10 - 11 | 1159 | 87 | |
| GenBank mRNA | BC086641.1 | - | 10 - 11 | 1159 | 6417957 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc44a5
Genomic Map