Assay Details
| Gene Symbol: Cul4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110142.1 | NP_001103612.1 | 2 - 3 | 198 | 61 | |
| NM_028288.5 | NP_082564.3 | 2 - 3 | 139 | 61 | ||
| GenBank mRNA | AK032701.1 | - | 2 - 3 | 140 | 61 | |
| AY330868.1 | - | 2 - 3 | 140 | 61 | ||
| BC004026.1 | - | 2 - 3 | 187 | 3494708 | 61 | |
| CJ072585.1 | - | 2 - 3 | 182 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC004026.1 transcripts.
Set Membership: |
|

Cul4b
Genomic Map