Assay Details
| Gene Symbol: Cul4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110142.1 | NP_001103612.1 | 7 - 8 | 1205 | 99 | |
| NM_028288.5 | NP_082564.3 | 7 - 8 | 1146 | 99 | ||
| GenBank mRNA | AB093259.1 | - | 4 - 5 | 467 | 99 | |
| AK012410.1 | - | 5 - 6 | 909 | 99 | ||
| AK032701.1 | - | 7 - 8 | 1147 | 99 | ||
| AK160998.1 | - | 5 - 6 | 916 | 99 | ||
| AK170632.1 | - | 4 - 5 | 1294 | 99 | ||
| AY330868.1 | - | 7 - 8 | 1147 | 99 | ||
| BC004026.1 | - | 7 - 8 | 1194 | 3494708 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC004026.1 transcripts.
Set Membership: |
|

Cul4b
Genomic Map