Assay Details
| Gene Symbol: Btg1-ps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034856.2 | NP_001030028.1 | 5 | 984 | 124 | |
| GenBank mRNA | AK132831.1 | - | 5 | 986 | 124 | |
| BC147535.1 | - | 2 | 574 | 9056526 | 124 | |
| BC147539.1 | - | 2 | 574 | 9056530 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996983 | NM_001034856.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996983 | AK132831.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664836 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996983 | NM_001034856.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996983 | AK132831.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03656765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32326001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32325953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14864221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15157626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14809751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14835259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14835260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14809748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14951855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14809758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14835264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14997597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32325992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32326011 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604160 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604163 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604161 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604162 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604159 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00013233.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013233.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013233.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013237.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013237.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013237.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013232.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013232.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013232.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013236.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013236.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013236.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013235.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013235.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013235.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013234.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013234.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013234.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013231.mm.1 | NM_001034856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013231.mm.1 | NM_001034856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013231.mm.1 | TC0X00002079.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 781868 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161824 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161824 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85702196-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161824 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161824 | - | - |

Btg1-ps2
Genomic Map