Assay Details
| Gene Symbol: Ppp2r2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205188.1 | NP_001192117.1 | 8 - 9 | 1354 | 72 | |
| NM_028032.3 | NP_082308.1 | 8 - 9 | 1354 | 72 | ||
| GenBank mRNA | AK010380.1 | - | 8 - 9 | 1319 | 72 | |
| AK090027.1 | - | 8 - 9 | 1340 | 72 | ||
| AK161507.1 | - | 8 - 9 | 1312 | 72 | ||
| AK161704.1 | - | 8 - 9 | 1303 | 72 | ||
| AK164497.1 | - | 8 - 9 | 1338 | 72 | ||
| AK167189.1 | - | 8 - 9 | 1343 | 72 | ||
| AK167530.1 | - | 8 - 9 | 1302 | 72 | ||
| AK220192.1 | - | 8 - 9 | 1307 | 72 | ||
| BC022916.1 | - | 9 - 10 | 1127 | 72 | ||
| BC038824.1 | - | 2 - 3 | 348 | 72 | ||
| BC054850.1 | - | 5 - 6 | 653 | 72 | ||
| BC065100.1 | - | 8 - 9 | 1327 | 30535680 | 72 | |
| BC080692.1 | - | 8 - 9 | 1277 | 30431716 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppp2r2a
Genomic Map