Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 11 - 12 | 1012 | 81 | |
| GenBank mRNA | AK131164.1 | - | 11 - 12 | 955 | 81 | |
| AK148355.1 | - | 11 - 12 | 1035 | 81 | ||
| AK158466.1 | - | 12 - 13 | 1258 | 81 | ||
| BC082805.1 | - | 11 - 12 | 932 | 81 | ||
| BC092223.1 | - | 11 - 12 | 930 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map