Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 13 - 14 | 1212 | 64 | |
| GenBank mRNA | AK131164.1 | - | 13 - 14 | 1155 | 64 | |
| AK148355.1 | - | 13 - 14 | 1235 | 64 | ||
| AK158466.1 | - | 14 - 15 | 1458 | 64 | ||
| BC082805.1 | - | 13 - 14 | 1132 | 64 | ||
| BC092223.1 | - | 13 - 14 | 1130 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map