Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 15 - 16 | 1372 | 80 | |
| GenBank mRNA | AK131164.1 | - | 15 - 16 | 1315 | 80 | |
| AK148355.1 | - | 15 - 16 | 1395 | 80 | ||
| AK158466.1 | - | 16 - 17 | 1618 | 80 | ||
| BC082805.1 | - | 15 - 16 | 1292 | 80 | ||
| BC092223.1 | - | 15 - 16 | 1290 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map