Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 17 - 18 | 1540 | 73 | |
| GenBank mRNA | AK131164.1 | - | 17 - 18 | 1483 | 73 | |
| AK148355.1 | - | 17 - 18 | 1563 | 73 | ||
| AK158466.1 | - | 18 - 19 | 1786 | 73 | ||
| BC036313.1 | - | 2 - 3 | 102 | 73 | ||
| BC082805.1 | - | 17 - 18 | 1460 | 73 | ||
| BC092223.1 | - | 17 - 18 | 1458 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map