Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 20 - 21 | 1781 | 73 | |
| GenBank mRNA | AK131164.1 | - | 20 - 21 | 1724 | 73 | |
| AK148355.1 | - | 20 - 21 | 1804 | 73 | ||
| AK158466.1 | - | 21 - 22 | 2027 | 73 | ||
| BC036313.1 | - | 5 - 6 | 343 | 73 | ||
| BC060252.1 | - | 1 - 2 | 321 | 73 | ||
| BC082805.1 | - | 20 - 21 | 1701 | 73 | ||
| BC092223.1 | - | 20 - 21 | 1699 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map