Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 3 - 4 | 335 | 71 | |
| GenBank mRNA | AK131164.1 | - | 3 - 4 | 337 | 71 | |
| AK148355.1 | - | 3 - 4 | 358 | 71 | ||
| AK158466.1 | - | 4 - 5 | 598 | 71 | ||
| BC082805.1 | - | 3 - 4 | 255 | 71 | ||
| BC092223.1 | - | 3 - 4 | 262 | 71 | ||
| CB248209.1 | - | 3 - 4 | 335 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map