Assay Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024645.1 | NP_001019816.1 | 5 - 6 | 511 | 103 | |
| GenBank mRNA | AK131164.1 | - | 5 - 6 | 513 | 103 | |
| AK148355.1 | - | 5 - 6 | 534 | 103 | ||
| BC082805.1 | - | 5 - 6 | 431 | 103 | ||
| BC092223.1 | - | 5 - 6 | 438 | 103 | ||
| CB248209.1 | - | 5 - 6 | 511 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc16b
Genomic Map