Assay Details
| Gene Symbol: Sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026474.5 | NP_080750.1 | 4 - 5 | 396 | 137 | |
| GenBank mRNA | AK007881.1 | - | 4 - 5 | 331 | 137 | |
| AK010711.1 | - | 3 - 4 | 365 | 137 | ||
| AK010830.1 | - | 4 - 5 | 313 | 137 | ||
| AK020433.1 | - | 4 - 5 | 315 | 137 | ||
| AK045153.1 | - | 4 - 5 | 335 | 137 | ||
| AK082328.1 | - | 4 - 5 | 398 | 137 | ||
| AK141416.1 | - | 332 | 137 | |||
| AK148838.1 | - | 4 - 5 | 310 | 137 | ||
| BC009167.1 | - | 4 - 5 | 313 | 2811924 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010711.1 transcripts.
Set Membership: |
|

Sugt1
Genomic Map