Assay Details
| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF269152.1 | - | 1 - 2 | 80 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF269152.1 transcripts.
Set Membership: |
|

Tm9sf1
Genomic Map