Assay Details
| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028780.3 | NP_083056.2 | 3 - 4 | 1406 | 55 | |
| GenBank mRNA | AF269152.1 | - | 3 - 4 | 1054 | 55 | |
| AK004754.2 | - | 3 - 4 | 1408 | 55 | ||
| AK090065.1 | - | 2 - 3 | 1401 | 55 | ||
| AK152476.1 | - | 3 - 4 | 1186 | 55 | ||
| AK152540.1 | - | 3 - 4 | 1186 | 55 | ||
| BC007187.1 | - | 2 - 3 | 711 | 55 | ||
| BC017617.1 | - | 2 - 3 | 1345 | 4923337 | 55 | |
More Information
Set Membership: |
|

Tm9sf1
Genomic Map