Assay Details
| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029417.1 | - | 1 - 2 | 79 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029417.1 transcripts.
Set Membership: |
|

Slc9b2
Genomic Map