Assay Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028946.3 | NP_083222.2 | 2 - 3 | 536 | 106 | |
| GenBank mRNA | AK007064.1 | - | 2 - 3 | 528 | 106 | |
| AK015318.1 | - | 2 - 3 | 503 | 106 | ||
| AK016883.1 | - | 2 - 3 | 525 | 106 | ||
| AK016917.1 | - | 2 - 3 | 516 | 106 | ||
| AK029525.1 | - | 2 - 3 | 537 | 106 | ||
| BC025002.1 | - | 1 - 2 | 458 | 106 | ||
| EU846100.1 | - | 1 - 2 | 67 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007064.1, AK015318.1, AK016883.1, AK016917.1, AK029525.1, BC025002.1, EU846100.1, NM_028946.3 transcripts.
Set Membership: |
|

Slc9b1
Genomic Map