Assay Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028946.3 | NP_083222.2 | 3 - 4 | 599 | 106 | |
| GenBank mRNA | AK007064.1 | - | 3 - 4 | 591 | 106 | |
| AK015318.1 | - | 3 - 4 | 566 | 106 | ||
| AK016883.1 | - | 3 - 4 | 588 | 106 | ||
| AK016917.1 | - | 3 - 4 | 579 | 106 | ||
| AK029525.1 | - | 3 - 4 | 600 | 106 | ||
| BC025002.1 | - | 2 - 3 | 521 | 106 | ||
| EU846100.1 | - | 2 - 3 | 130 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9b1
Genomic Map