Assay Details
| Gene Symbol: Dnajb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033155.1 | NP_001028327.1 | 7 - 8 | 1259 | 153 | |
| GenBank mRNA | AK017735.1 | - | 7 - 8 | 1120 | 153 | |
| BC117717.1 | - | 7 - 8 | 1060 | 40088109 | 153 | |
| BC117718.1 | - | 7 - 8 | 1060 | 40088110 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137475 | BC117717.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137475 | BC117718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137475 | NM_001033155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137475 | AK017735.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524092 | AK029214.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441920 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289045 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80122 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143628 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137475 | BC117717.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137475 | BC117718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137475 | NM_001033155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137475 | AK017735.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143628 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05089289 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00768061 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05089262 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05089233 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05089246 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05089277 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02822699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22565671 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10496389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10496392 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430561_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10496387 | - | - |
| Mouse Genome 430 2.0 Array | 1430561_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430561_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70604_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70604_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300011275.mm.1 | NM_001033155.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300011275.mm.1 | NM_001033155.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300011275.mm.1 | TC0300001399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300011276.mm.1 | NM_001033155.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300011276.mm.1 | NM_001033155.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300011276.mm.1 | TC0300001399.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430561_at | - | - |
| Mouse Expression Set 430 | 1430561_PM_at | - | - |
| Murine Genome U74v2 Set | 168478_s_at | - | - |
| Murine Genome U74v2 Set | 168478_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430561_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430561_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 704093 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2661062 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2661062 | - | - |

Dnajb14
Genomic Map