Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 6 - 7 | 667 | 60 | |
| NM_173763.4 | NP_776124.1 | 6 - 7 | 671 | 60 | ||
| GenBank mRNA | AF363737.1 | - | 6 - 7 | 641 | 60 | |
| AK049569.1 | - | 6 - 7 | 640 | 60 | ||
| AK145623.1 | - | 6 - 7 | 665 | 60 | ||
| BC037135.1 | - | 2 - 3 | 248 | 60 | ||
| BC085287.1 | - | 7 - 8 | 764 | 60 | ||
| BC131942.1 | - | 5 - 6 | 456 | 40130219 | 60 | |
| BC132615.1 | - | 5 - 6 | 456 | 40130892 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl2
Genomic Map