Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 9 - 10 | 996 | 60 | |
| NM_173763.4 | NP_776124.1 | 9 - 10 | 1000 | 60 | ||
| GenBank mRNA | AF363737.1 | - | 9 - 10 | 60 | ||
| AK049569.1 | - | 9 - 10 | 969 | 60 | ||
| AK145623.1 | - | 9 - 10 | 994 | 60 | ||
| BC037135.1 | - | 5 - 6 | 60 | |||
| BC085287.1 | - | 10 - 11 | 60 | |||
| BC131942.1 | - | 8 - 9 | 40130219 | 60 | ||
| BC132615.1 | - | 8 - 9 | 40130892 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl2
Genomic Map