Assay Details
| Gene Symbol: Znhit6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081094.1 | NP_001074563.1 | 3 - 4 | 963 | 66 | |
| GenBank mRNA | AK148729.1 | - | 3 - 4 | 940 | 66 | |
| AK167835.1 | - | 4 - 5 | 385 | 66 | ||
| AK168844.1 | - | 4 - 5 | 379 | 66 | ||
| BC147498.1 | - | 3 - 4 | 849 | 9056360 | 66 | |
| BC150966.1 | - | 3 - 4 | 849 | 9087878 | 66 | |
| BC150967.1 | - | 3 - 4 | 849 | 9087879 | 66 | |
| BC151198.1 | - | 3 - 4 | 849 | 9088086 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Znhit6
Genomic Map