Assay Details
| Gene Symbol: Znhit6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081094.1 | NP_001074563.1 | 7 - 8 | 1303 | 112 | |
| GenBank mRNA | AK010555.1 | - | 6 - 7 | 648 | 112 | |
| AK148729.1 | - | 7 - 8 | 1280 | 112 | ||
| AK167835.1 | - | 8 - 9 | 726 | 112 | ||
| AK168844.1 | - | 8 - 9 | 719 | 112 | ||
| BC147498.1 | - | 7 - 8 | 1189 | 9056360 | 112 | |
| BC150966.1 | - | 7 - 8 | 1189 | 9087878 | 112 | |
| BC150967.1 | - | 7 - 8 | 1189 | 9087879 | 112 | |
| BC151198.1 | - | 7 - 8 | 1189 | 9088086 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Znhit6
Genomic Map