Assay Details
| Gene Symbol: Rhcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019799.3 | NP_062773.2 | 8 - 9 | 1372 | 60 | |
| GenBank mRNA | AF193810.1 | - | 8 - 9 | 1363 | 60 | |
| AK076757.1 | - | 8 - 9 | 1372 | 60 | ||
| AY254686.1 | - | 8 - 9 | 1247 | 60 | ||
| BC119045.1 | - | 8 - 9 | 1331 | 8733794 | 60 | |
| BC137652.1 | - | 8 - 9 | 1313 | 8860671 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhcg
Genomic Map