Assay Details
| Gene Symbol: Picalm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252520.1 | NP_001239449.1 | 15 - 16 | 1975 | 106 | |
| NM_001252521.1 | NP_001239450.1 | 15 - 16 | 1990 | 106 | ||
| NM_001252522.1 | NP_001239451.1 | 14 - 15 | 1840 | 106 | ||
| NM_001252523.1 | NP_001239452.1 | 14 - 15 | 1825 | 106 | ||
| NM_001252524.1 | NP_001239453.1 | 14 - 15 | 1825 | 106 | ||
| NM_146194.4 | NP_666306.2 | 15 - 16 | 1990 | 106 | ||
| GenBank mRNA | AK085472.2 | - | 11 - 12 | 1059 | 106 | |
| AK144960.1 | - | 15 - 16 | 1731 | 106 | ||
| AK162360.1 | - | 15 - 16 | 1668 | 106 | ||
| AK165066.1 | - | 9 - 10 | 937 | 106 | ||
| AK170373.1 | - | 14 - 15 | 1845 | 106 | ||
| AK220291.1 | - | 15 - 16 | 1691 | 106 | ||
| AY206701.1 | - | 15 - 16 | 1674 | 106 | ||
| BC011470.1 | - | 15 - 16 | 1685 | 2651109 | 106 | |
| BC021491.1 | - | 14 - 15 | 1774 | 5345558 | 106 | |
| BC023843.1 | - | 15 - 16 | 1738 | 5346041 | 106 | |
| BC025566.1 | - | 14 - 15 | 1863 | 5322606 | 106 | |
| BC027116.1 | - | 12 - 13 | 1286 | 106 | ||
| BC057683.1 | - | 14 - 15 | 1776 | 3671962 | 106 | |
More Information
Set Membership: |
|

Picalm
Genomic Map