Assay Details
| Gene Symbol: Blm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042527.2 | NP_001035992.1 | 10 - 11 | 2413 | 124 | |
| NM_007550.4 | NP_031576.4 | 10 - 11 | 2684 | 124 | ||
| GenBank mRNA | AB008674.1 | - | 10 - 11 | 2674 | 124 | |
| AK137022.1 | - | 10 - 11 | 2396 | 124 | ||
| AK157898.1 | - | 10 - 11 | 2414 | 124 | ||
| Z98263.1 | - | 10 - 11 | 2391 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Blm
Genomic Map