| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700030265.mm.1 |
NM_001042527.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030265.mm.1 |
NM_001042527.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030265.mm.1 |
NM_007550.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030265.mm.1 |
NM_007550.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030265.mm.1 |
TC0700004655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030266.mm.1 |
NM_001042527.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030266.mm.1 |
NM_001042527.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030266.mm.1 |
NM_007550.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030266.mm.1 |
NM_007550.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030266.mm.1 |
TC0700004655.mm.1 |
MTA TC |