Assay Details
| Gene Symbol: Cenpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014996.2 | NP_001014996.2 | 10 - 11 | 3394 | 92 | |
| GenBank mRNA | AK077045.1 | - | 10 - 11 | 3377 | 92 | |
| BC092399.1 | - | 10 - 11 | 3380 | 30549055 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpj
Genomic Map