Assay Details
| Gene Symbol: Etnppl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163587.1 | NP_001157059.1 | 10 - 11 | 1164 | 87 | |
| NM_027907.3 | NP_082183.2 | 9 - 10 | 1202 | 87 | ||
| GenBank mRNA | AK005060.1 | - | 9 - 10 | 1203 | 87 | |
| AK049937.1 | - | 10 - 11 | 1164 | 87 | ||
| BC043680.1 | - | 9 - 10 | 1189 | 5101568 | 87 | |
| BC058592.1 | - | 9 - 10 | 1164 | 5363337 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etnppl
Genomic Map