Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300010714.mm.1 |
NM_001163587.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010714.mm.1 |
NM_001163587.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010714.mm.1 |
NM_027907.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010714.mm.1 |
NM_027907.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010714.mm.1 |
TC0300001322.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010713.mm.1 |
NM_001163587.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010713.mm.1 |
NM_001163587.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010713.mm.1 |
NM_027907.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010713.mm.1 |
NM_027907.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010713.mm.1 |
TC0300001322.mm.1 |
MTA TC |