Assay Details
| Gene Symbol: Etnppl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163587.1 | NP_001157059.1 | 7 - 8 | 704 | 85 | |
| NM_027907.3 | NP_082183.2 | 6 - 7 | 742 | 85 | ||
| GenBank mRNA | AK005060.1 | - | 6 - 7 | 743 | 85 | |
| AK049937.1 | - | 7 - 8 | 704 | 85 | ||
| BC043680.1 | - | 6 - 7 | 729 | 5101568 | 85 | |
| BC058592.1 | - | 6 - 7 | 704 | 5363337 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etnppl
Genomic Map