Assay Details
| Gene Symbol: Dnttip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153806.1 | NP_722501.1 | 2 - 3 | 1682 | 68 | |
| GenBank mRNA | AK133904.1 | - | 2 - 3 | 1701 | 68 | |
| AK163260.1 | - | 1 - 2 | 299 | 68 | ||
| AK168536.1 | - | 2 - 3 | 1701 | 68 | ||
| BC028305.1 | - | 2 - 3 | 1682 | 2609541 | 68 | |
More Information
Set Membership: |
|

Dnttip2
Genomic Map