Assay Details
| Gene Symbol: Pde10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290707.1 | NP_001277636.1 | 2 - 3 | 450 | 89 | |
| GenBank mRNA | AK039249.1 | - | 2 - 3 | 450 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039249.1 transcripts.
Set Membership: |
|

Pde10a
Genomic Map