Assay Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145514.5 | NP_663489.4 | 11 - 12 | 2129 | 140 | |
| GenBank mRNA | AK075743.1 | - | 11 - 12 | 1521 | 140 | |
| AK082326.1 | - | 12 - 13 | 1424 | 140 | ||
| AK213047.1 | - | 2 - 3 | 121 | 140 | ||
| AK218758.1 | - | 2 - 3 | 121 | 140 | ||
| BC020044.1 | - | 7 - 8 | 874 | 140 | ||
| BC058601.1 | - | 9 - 10 | 1125 | 140 | ||
| BC094233.1 | - | 8 - 9 | 1013 | 140 | ||
| BC139336.1 | - | 11 - 12 | 1291 | 8862358 | 140 | |
| BC139337.1 | - | 11 - 12 | 1291 | 8862359 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr26
Genomic Map