Assay Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145514.5 | NP_663489.4 | 12 - 13 | 2257 | 80 | |
| GenBank mRNA | AK075743.1 | - | 12 - 13 | 1649 | 80 | |
| AK082326.1 | - | 13 - 14 | 1552 | 80 | ||
| AK167017.1 | - | 1 - 2 | 66 | 80 | ||
| AK167116.1 | - | 1 - 2 | 66 | 80 | ||
| BC020044.1 | - | 8 - 9 | 1002 | 80 | ||
| BC058601.1 | - | 10 - 11 | 1253 | 80 | ||
| BC094233.1 | - | 9 - 10 | 1141 | 80 | ||
| BC139336.1 | - | 12 - 13 | 1419 | 8862358 | 80 | |
| BC139337.1 | - | 12 - 13 | 1419 | 8862359 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr26
Genomic Map