Assay Details
| Gene Symbol: Nsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198654.3 | NP_941056.3 | 2 - 3 | 327 | 89 | |
| GenBank mRNA | BC029086.1 | - | 2 - 3 | 317 | 3156934 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Nsl1
Genomic Map