Assay Details
| Gene Symbol: Iars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198653.2 | NP_941055.1 | 20 - 21 | 2668 | 80 | |
| GenBank mRNA | AK036182.1 | - | 20 - 21 | 2636 | 80 | |
| AK043590.1 | - | 20 - 21 | 2670 | 80 | ||
| AK048653.1 | - | 20 - 21 | 2669 | 80 | ||
| AK050358.1 | - | 20 - 21 | 2651 | 80 | ||
| AK051778.1 | - | 20 - 21 | 2628 | 80 | ||
| AK078183.1 | - | 20 - 21 | 2670 | 80 | ||
| AK085714.1 | - | 20 - 21 | 2666 | 80 | ||
| AK088965.1 | - | 20 - 21 | 2629 | 80 | ||
| AK166832.1 | - | 20 - 21 | 2651 | 80 | ||
| BC027432.1 | - | 10 - 11 | 1126 | 80 | ||
| BC049238.1 | - | 4 - 5 | 407 | 80 | ||
| BC052403.1 | - | 20 - 21 | 2645 | 5715723 | 80 | |
More Information
Set Membership: |
|

Iars2
Genomic Map