Assay Details
| Gene Symbol: Iars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198653.2 | NP_941055.1 | 21 - 22 | 2862 | 100 | |
| GenBank mRNA | AK036182.1 | - | 21 - 22 | 2830 | 100 | |
| AK043590.1 | - | 21 - 22 | 2864 | 100 | ||
| AK048653.1 | - | 21 - 22 | 2863 | 100 | ||
| AK050358.1 | - | 21 - 22 | 2845 | 100 | ||
| AK051778.1 | - | 21 - 22 | 2822 | 100 | ||
| AK078183.1 | - | 21 - 22 | 2864 | 100 | ||
| AK085714.1 | - | 21 - 22 | 2860 | 100 | ||
| AK088965.1 | - | 21 - 22 | 2823 | 100 | ||
| AK166832.1 | - | 21 - 22 | 2845 | 100 | ||
| BC027432.1 | - | 11 - 12 | 1320 | 100 | ||
| BC049238.1 | - | 5 - 6 | 601 | 100 | ||
| BC052403.1 | - | 21 - 22 | 2839 | 5715723 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iars2
Genomic Map