Assay Details
| Gene Symbol: Ahctf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026375.2 | NP_080651.2 | 21 - 22 | 3331 | 149 | |
| GenBank mRNA | AB059278.1 | - | 21 - 22 | 2987 | 149 | |
| BC138343.1 | - | 21 - 22 | 2974 | 8861363 | 149 | |
| BC138344.1 | - | 21 - 22 | 2974 | 8861364 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahctf1
Genomic Map